Order Entry
Poland
ContactUsLinkComponent
2-(4-Fluorophenyl)-2-propanol 97%
Irritant
2-(4-Fluorophenyl)-2-propanol 97%
Catalog # APOSPC4174E-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(4-Fluorophenyl)-2-propanol 97%
Catalog # APOSPC4174E-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.18 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:402-41-5
Formula:C₉H₁₁FO
MDL Number:MFCD00059919