1-(Difluoromethyl)-5-methyl-1H-pyrazole
Catalog # APOSPC410309-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:132.11 g/mol
Storage Temperature:Ambient
CAS Number:1004096-15-4
Formula:C₅H₆F₂N₂
MDL Number:MFCD04968779