
2-(3-Trifluoromethyl-α-toluenesulphonyl)thioacetamide 97%
Catalog # APOSPC4005-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:165…171 °C
:GHS07
Storage Temperature:Ambient
CAS Number:175276-83-2
Formula:C₁₀H₁₀F₃NO₂S₂
MDL Number:MFCD00052940