
5-Fluoro-2-toluoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21900-39-0
:
Formula:C₈H₆ClFO
MDL Number:MFCD00236681
:172.59 g/mol
Boiling Pt:108 °C/24mmHg
Density:1.27 g/cm³ (25 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together