Order Entry
Poland
ContactUsLinkComponent
2,3-Dichloro-1,1,1,4,4,4-hexafluorobutane 97%
Irritant
2,3-Dichloro-1,1,1,4,4,4-hexafluorobutane 97%
Catalog # APOSPC3779-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Dichloro-1,1,1,4,4,4-hexafluorobutane 97%
Catalog # APOSPC3779-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:83…84 °C
 :234.96 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:384-54-3
Formula:C₄H₂Cl₂F₆
MDL Number:MFCD02093296