Order Entry
Poland
ContactUsLinkComponent
2-Bromo-N-ethy-4-(trifluoromethoxy)benzenesulphonamide
2-Bromo-N-ethy-4-(trifluoromethoxy)benzenesulphonamide
Catalog # APOSPC3492-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-N-ethy-4-(trifluoromethoxy)benzenesulphonamide
Catalog # APOSPC3492-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:957062-74-7
Formula:C₉H₉BrF₃NO₃S
MDL Number:MFCD09878394