Order Entry
Poland
ContactUsLinkComponent
3,3,3-Trifluoropropane-1-sulphonyl chloride 97%
Corrosive
3,3,3-Trifluoropropane-1-sulphonyl chloride 97%
Catalog # APOSPC3467-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3,3,3-Trifluoropropane-1-sulphonyl chloride 97%
Catalog # APOSPC3467-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :196.58 g/mol
Boiling Pt:77…78 °C/16mmHg
Density:1.557 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
 :GHS05
CAS Number:845866-80-0
Formula:C₃H₄ClF₃O₂S
MDL Number:MFCD06248898