3-Bromo-N,N-dipropyl-5-(trifluoromethyl)benzenesulphonamide
Catalog # APOSPC3448-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:957062-78-1
Formula:C₁₃H₁₇BrF₃NO₂S
MDL Number:MFCD09878371