Order Entry
Poland
ContactUsLinkComponent
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine 98%
Toxic
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine 98%
Catalog # APOSPC3352-25G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine 98%
Catalog # APOSPC3352-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :260.44 g/mol
Melting Pt:28…32 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:71701-92-3
Formula:C₆H₂BrClF₃N
MDL Number:MFCD09878432