Order Entry
Poland
ContactUsLinkComponent
1H,1H-Perfluorohexyl-p-toluenesulphonate 97%
1H,1H-Perfluorohexyl-p-toluenesulphonate 97%
Catalog # APOSPC3346-1G
Supplier:  Apollo Scientific
CAS Number:  
1H,1H-Perfluorohexyl-p-toluenesulphonate 97%
Catalog # APOSPC3346-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :454.26 g/mol
Flash Pt:>110 °C
Storage Temperature:Ambient
CAS Number:355-77-1
Formula:C₁₃H₉F₁₁O₃S
MDL Number:MFCD04038342