Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
APOSPC32296-1G
:
Apollo Scientific
:
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
APOSPC32296-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
214.15 g/mol
:
GHS07
:
Ambient
:
2251-79-8
:
MFCD00052525
:
C₈H₅F₃N₄
:
MFCD06248886