
2-Bromo-5-fluorophenyl isothiocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1027513-65-0
:
Formula:C₇H₃BrFNS
MDL Number:MFCD00276607
ADR:8,III
:232.08 g/mol
UN:1760
:GHS05
Storage Temperature:Refrigerator
Frequently Bought Together