
3-Bromo-5-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54962-75-3
:
Formula:C₇H₅BrF₃N
MDL Number:MFCD00236205
:240.02 g/mol
Boiling Pt:220…223 °C
Density:1.697 g/cm³ (25 °C)
Flash Pt:107 °C
EINECS:259-412-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together