
1H,1H,11H-Eicosafluoro-1-undecanol 85%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:307-70-0
:
Formula:C₁₁H₄F₂₀O
MDL Number:MFCD00039628
:532.12 g/mol
Boiling Pt:180…181 °C/200mmHg
Melting Pt:95…97 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together