Order Entry
Poland
ContactUsLinkComponent
2-Chloro-6-(trifluoromethyl)benzonitrile 98%
2-Chloro-6-(trifluoromethyl)benzonitrile 98%
Catalog # APOSPC303105-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-6-(trifluoromethyl)benzonitrile 98%
Catalog # APOSPC303105-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.57
Melting Pt:52…54 °C
Storage Temperature:Ambient
CAS Number:129604-28-0
Formula:C₈H₃ClF₃N
MDL Number:MFCD03412191