Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-5-(pentafluorothio)cinnamic acid
3-Fluoro-5-(pentafluorothio)cinnamic acid 97%
3-Fluoro-5-(pentafluorothio)cinnamic acid 97%
APOSPC302845-1G
:
Apollo Scientific
:
3-Fluoro-5-(pentafluorothio)cinnamic acid 97%
APOSPC302845-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
292.20 g/mol
:
Ambient
:
C₉H₆F₆O₂S
:
MFCD16652465