Order Entry
Poland
ContactUsLinkComponent
3-Methoxy-2-(trifluoromethyl)benzylamine 98%
Corrosive
3-Methoxy-2-(trifluoromethyl)benzylamine 98%
Catalog # APOSPC302730-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3-Methoxy-2-(trifluoromethyl)benzylamine 98%
Catalog # APOSPC302730-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.18 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
CAS Number:1261647-71-5
Formula:C₉H₁₀F₃NO
MDL Number:MFCD18398592