Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(4-Ethoxy-2,3-difluorophenyl)acetic acid
(4-Ethoxy-2,3-difluorophenyl)acetic acid 97%
(4-Ethoxy-2,3-difluorophenyl)acetic acid 97%
APOSPC302583-1G
:
Apollo Scientific
:
(4-Ethoxy-2,3-difluorophenyl)acetic acid 97%
APOSPC302583-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
145…148 °C
:
Ambient
:
1017779-62-2
:
C₁₀H₁₀F₂O₃
:
MFCD09258707