
4-(4-Morpholinyl)-2-(trifluoromethyl)aniline
Catalog # APOSPC300734-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:246.23 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:78335-26-9
Formula:C₁₁H₁₃F₃N₂O
MDL Number:MFCD01436021