2-Chloro-4-(trifluoromethyl)benzenesulphonamide
Catalog # APOSPC2976-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.64
Storage Temperature:Ambient
CAS Number:146533-47-3
Formula:C₇H₅ClF₃NO₂S
MDL Number:MFCD00179384