
2,4-Difluoro-6-nitroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:364-30-7
:
Formula:C₆H₄F₂N₂O₂
MDL Number:MFCD00034058
:174.11 g/mol
Melting Pt:85…87 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together