
cis-1,1,2,2,3,4-Hexafluorocyclobutane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22819-47-2
:
Formula:C₄H₂F₆
MDL Number:MFCD03094205
:164.05 g/mol
Boiling Pt:63 °C
Density:1.778 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together