
5-Fluoro-m-anisaldehyde 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:699016-24-5
:
Formula:C₈H₇FO₂
MDL Number:MFCD08741376
:154.14 g/mol
Melting Pt:24…29 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together