

1,3-Dichloro-2-fluorobenzene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2268-05-5
:
Formula:C₆H₃Cl₂F
MDL Number:MFCD00010307
:164.99 g/mol
Boiling Pt:168…169 °C
Melting Pt:36…38 °C
Flash Pt:60 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1,II
:GHS02
:GHS07
Frequently Bought Together