(Aza((4-fluorophenyl)amino)methylene)methane-1,1-dicarbonitrile
Catalog # APOSPC210059-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:188.17 g/mol
Storage Temperature:Ambient
CAS Number:94853-74-4
Formula:C₉H₅FN₄
MDL Number:MFCD00170788