Order Entry
Poland
ContactUsLinkComponent
Copper(II) tetrafluoroborate hexahydrate 98%
Corrosive
Copper(II) tetrafluoroborate hexahydrate 98%
Catalog # APOSPC2095-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Copper(II) tetrafluoroborate hexahydrate 98%
Catalog # APOSPC2095-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :345.243 g/mol
Storage Temperature:Ambient
EINECS:253-959-4
UN:3260
ADR:8,III
 :GHS05
CAS Number:72259-10-0
Formula:B₂CuF₈H₁₂O₆
MDL Number:MFCD00150223