5-(1,3-Dioxan-2-yl)-2-fluorobenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:218301-23-6
:
Formula:C₁₁H₁₀FNO₂
MDL Number:MFCD20921611
:207.21 g/mol
Melting Pt:45…47 °C
Storage Temperature:Ambient
Frequently Bought Together