Order Entry
Poland
ContactUsLinkComponent
2-{3-[2-Amino-5-(trifluoromethyl)-3-pyridinyl]-2-propynyl}-1H-isoindole-1,3(2H)-dione
2-{3-[2-Amino-5-(trifluoromethyl)-3-pyridinyl]-2-propynyl}-1H-isoindole-1,3(2H)-dione
Catalog # APOSPC200309-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-{3-[2-Amino-5-(trifluoromethyl)-3-pyridinyl]-2-propynyl}-1H-isoindole-1,3(2H)-dione
Catalog # APOSPC200309-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :345.28 g/mol
Melting Pt:186…188 °C
Storage Temperature:Ambient
CAS Number:1221792-28-4
Formula:C₁₇H₁₀F₃N₃O₂
MDL Number:MFCD14584841