4-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1211579-79-1
:
Formula:C₇H₅F₃N₄
MDL Number:MFCD18261252
:202.14 g/mol
Melting Pt:170…172 °C
Storage Temperature:Ambient
Frequently Bought Together