5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1036027-54-9
:
Formula:C₈H₅F₃N₂
MDL Number:MFCD06659680
:186.1300 g/mol
Melting Pt:166 °C
Storage Temperature:Ambient
Frequently Bought Together