Order Entry
Poland
ContactUsLinkComponent
2,5-Dichloro-4-(difluoromethyl)pyridine 97%
Irritant
2,5-Dichloro-4-(difluoromethyl)pyridine 97%
Catalog # APOSPC200124-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,5-Dichloro-4-(difluoromethyl)pyridine 97%
Catalog # APOSPC200124-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1374659-30-9
Formula:C₆H₃Cl₂F₂N
MDL Number:MFCD22199291