Order Entry
Poland
ContactUsLinkComponent
5-Bromo-2-fluorocinnamic acid 98%
5-Bromo-2-fluorocinnamic acid 98%
Catalog # APOSPC1423T-25G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-2-fluorocinnamic acid 98%
Catalog # APOSPC1423T-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :245.05 g/mol
Melting Pt:194…196 °C
Storage Temperature:Ambient
CAS Number:202865-71-2
Formula:C₉H₆BrFO₂
MDL Number:MFCD00143427