
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:486460-21-3
:
Formula:C₆H₇F₃N₄
MDL Number:MFCD08460920
:192.145 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together