
Ammonium hexafluorogermanate(IV) 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16962-47-3
:
Formula:H₈F₆GeN₂
MDL Number:MFCD00010880
:222.68 g/mol
Melting Pt:380 °C
Storage Temperature:Ambient
UN:3288
ADR:6.1,III
:GHS07
Frequently Bought Together