
2-Bromo-5-(trifluorometylo)anilina 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:454-79-5
:
Formula:C₇H₅BrF₃N
MDL Number:MFCD00007637
Boiling Pt:81…82 °C/5mmHg
Density:1,675 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Temperatura otoczenia
EINECS:207-227-6
UN:3267
ADR:8,II
:240.02 g/mol
:GHS05
Frequently Bought Together