

3-Fluorophenyl isothiocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:404-72-8
:
Formula:C₇H₄FNS
MDL Number:MFCD00004804
:153.18 g/mol
Boiling Pt:227 °C
Density:1.27 g/cm³ (25 °C)
Flash Pt:85 °C
Storage Temperature:Refrigerator
EINECS:206-967-7
UN:2922
ADR:8+6.1,III
:GHS05
:GHS07
Frequently Bought Together