
2,6-Dichloro-3-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:55304-75-1
:
Formula:C₆H₂Cl₂F₃N
MDL Number:MFCD00042245
Boiling Pt:194…196 °C
Melting Pt:44383 °C
Density:2.008 g/cm³ (25 °C)
Flash Pt:103 °C
EINECS:259-585-8
ADR:6.1,III
:215.99 g/mol
UN:2810
:GHS06
Storage Temperature:Ambient
Frequently Bought Together