N-(2,6-Difluorophenyl)-1-hydrazinecarboxamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:171277-87-5
:
Formula:C₇H₇F₂N₃O
MDL Number:MFCD03791164
:187.15 g/mol
Melting Pt:161…163 °C
Storage Temperature:Ambient
Frequently Bought Together