


4-(Trifluoromethyl)anilinium chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:90774-69-9
:
Formula:C₇H₇ClF₃N
MDL Number:MFCD00007897
:197.59
Melting Pt:200 °C
Storage Temperature:Refrigerator
EINECS:292-679-7
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Frequently Bought Together