
3-((Difluoromethyl)thio)benzoyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261944-16-5
:
Formula:C₈H₅ClF₂OS
:222.64
Boiling Pt:85…88 °C/8mmHg
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
Frequently Bought Together