
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 97%
Catalog # APOSOR9929-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:167.16 g/mol
Melting Pt:283 °C
:GHS07
Storage Temperature:Ambient
CAS Number:253870-02-9
Formula:C8H9NO3
MDL Number:MFCD06202342