Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl-3-(isopropyl)carbazate
tert-Butyl-3-(isopropyl)carbazate 98%
tert-Butyl-3-(isopropyl)carbazate 98%
APOSOR965099-5G
:
Apollo Scientific
:
tert-Butyl-3-(isopropyl)carbazate 98%
APOSOR965099-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
174.24 g/mol
:
Ambient
:
16689-35-3
:
C₈H₁₈N₂O₂
:
MFCD14635748