1-(2-Methoxyethyl)-1H-pyrazole-4-carbaldehyde 95%
Catalog # APOSOR948690-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154.17 g/mol
Storage Temperature:Ambient
CAS Number:304693-70-7
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD13248693