Order Entry
Poland
ContactUsLinkComponent
4,5-Dibromo-1-tritylimidazole 98%
4,5-Dibromo-1-tritylimidazole 98%
Catalog # APOSOR9417-1G
Supplier:  Apollo Scientific
CAS Number:  
4,5-Dibromo-1-tritylimidazole 98%
Catalog # APOSOR9417-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:223…225 °C
Storage Temperature:Refrigerator
CAS Number:112517-23-4
Formula:C₂₂H₁₆Br₂N₂
MDL Number:MFCD08056373