Order Entry
Poland
ContactUsLinkComponent
4-Chloro-2-methylthieno[2,3-d]pyrimidine 95%
4-Chloro-2-methylthieno[2,3-d]pyrimidine 95%
Catalog # APOSOR932569-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2-methylthieno[2,3-d]pyrimidine 95%
Catalog # APOSOR932569-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :184.65 g/mol
Storage Temperature:Ambient
CAS Number:56843-79-9
Formula:C₇H₅ClN₂S
MDL Number:MFCD03028468