Order Entry
Poland
ContactUsLinkComponent
7-Bromothieno[3,2-d]pyrimidine 98%
Irritant
7-Bromothieno[3,2-d]pyrimidine 98%
Catalog # APOSOR930033-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
7-Bromothieno[3,2-d]pyrimidine 98%
Catalog # APOSOR930033-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :215.08 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:21586-25-4
Formula:C₆H₃BrN₂S
MDL Number:MFCD11520387