Order Entry
Poland
ContactUsLinkComponent
ɑ-Methylene-γ-butyrolactone 95%
ɑ-Methylene-γ-butyrolactone 95%
Catalog # APOSOR928542-5G
Supplier:  Apollo Scientific
CAS Number:  
ɑ-Methylene-γ-butyrolactone 95%
Catalog # APOSOR928542-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :98.10 g/mol
Density:1.12 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:547-65-9
Formula:C₅H₆O₂
MDL Number:MFCD00005407