5-Chloroisoindoline-1,3-dione 95%
Catalog # APOSOR926828-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:181.5759 g/mol
Storage Temperature:Ambient
CAS Number:7147-90-2
Formula:C8H4ClNO2
MDL Number:MFCD08703319