Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-2-methylpropane
1-Bromo-2-methylpropane
1-Bromo-2-methylpropane
1-Bromo-2-methylpropane
APOSOR924726-1KG
:
Apollo Scientific
:
1-Bromo-2-methylpropane
APOSOR924726-1KG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 kg
Seq:
4024
:
:
137.02 g/mol
:
90…92 °C
:
–118 °C
:
1.255 g/cm³ (25 °C)
:
18 °C
:
2342
:
3,II
:
GHS02
:
GHS07
:
Ambient
:
78-77-3
:
C₄H₉Br
:
MFCD00000217