Order Entry
Poland
ContactUsLinkComponent
cis-2,6-Dimethylpiperidine 95%
cis-2,6-Dimethylpiperidine 95%
Catalog # APOSOR923199-25G
Supplier:  Apollo Scientific
CAS Number:  
cis-2,6-Dimethylpiperidine 95%
Catalog # APOSOR923199-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :113.20 g/mol
Storage Temperature:Ambient
CAS Number:766-17-6
Formula:C₇H₁₅N
MDL Number:MFCD00066304